1-Benzyl-4-hydroxypiperidin ≥97%

Lieferant: ABCR
Achtung

Synonyme: 1-Benzylpiperidin-4-ol

10932
ABCR10932EA 0 EUR
ABCR10932
1-Benzyl-4-hydroxypiperidin ≥97%
1-Benzyl-4-hydroxypiperidin

Purity: 97% CAS No.: 4727-72-4 Molecular Formula: C12H17NO Formula Weight: 191.27 Melting Point: 60-64° Boiling Point: 121-123°/0.7mm MDL No.: MFCD00006503

Formel: C₁₂H₁₇NO
Molecular Weight: 191,27 g/mol
Siedepunkt: 121…123 °C (0,7 mmHg)
Schmelzpunkt: 60…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006503
CAS-Nummer: 4727-72-4
EINECS: 225-226-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR