D(-)-Penicillamin 98%

Lieferant: abcr
Achtung

110575.0001
ABCR110575.0001EA 0 EUR
ABCR110575.0001
D(-)-Penicillamin 98%
D(-)-Penicillamin
Formel: C₅H₁₁NO₂S
Lagertemperatur: Kühlschrank
MDL: MFCD00064302
CAS-Nummer: 52-67-5
EINECS: 200-148-8
Merck Index: 14,7088

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR