2-Amino-3,5-dibrompyridin
Lieferant: ABCR
|
Achtung
|
Synonyme:
3,5-Dibrompyridin-2-amin, 3,5-Dibrom-2-pyridylamin
12706
ABCR12706EA
0
EUR
ABCR12706
2-Amino-3,5-dibrompyridin
2-Amino-3,5-dibrompyridin
Formel:
C₅H₄Br₂N₂ Molecular Weight: 251,91 g/mol Siedepunkt: 254 °C (1013 hPa) Schmelzpunkt: 104 °C |
MDL:
MFCD00038041 CAS-Nummer: 35486-42-1 EINECS: 252-590-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...