(4-Hydroxy-3-nitrophenyl)essigsäure ≥99%

Lieferant: ABCR
Achtung

Synonyme: 2-(4-Hydroxy-3-nitrophenyl)acetic acid

13858
ABCR13858EA 0 EUR
ABCR13858
(4-Hydroxy-3-nitrophenyl)essigsäure ≥99%
(4-Hydroxy-3-nitrophenyl)essigsäure
Formel: (C₆H₃(OH)(NO₂))CH₂COOH
Molecular Weight: 197,15 g/mol
Schmelzpunkt: 146…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007122
CAS-Nummer: 10463-20-4
EINECS: 233-948-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR