1-Methoxy-2,2,2-trifluorethanol

Lieferant: ABCR

Synonyme: Trifluoracetaldehyd-Methylhalbacetal, Trifluoracetaldehydmethylhemiacetal

18369.0025
ABCR18369.0025EA 0 EUR
ABCR18369.0025
1-Methoxy-2,2,2-trifluorethanol
1-Methoxy-2,2,2-trifluoroethanol
Formel: C₃H₅F₃O₂
Molecular Weight: 130,07 g/mol
MDL: MFCD00013572
CAS-Nummer: 431-46-9
EINECS: 207-072-4
UN: 3271
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR