3-(Trifluormethoxy)benzoylchlorid

Lieferant: ABCR

20084.0001
ABCR20084.0001EA 0 EUR
ABCR20084.0001
3-(Trifluormethoxy)benzoylchlorid
3-(Trifluormethoxy)benzoylchlorid
Formel: C₈H₄ClF₃O₂
Molecular Weight: 224,57 g/mol
MDL: MFCD00061268
CAS-Nummer: 86270-03-3
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR