3-Ethoxy-2-thenoesäure

Lieferant: ABCR

Synonyme: 3-Ethoxythiophene-2-carboxylic acid

20296.0001
ABCR20296.0001EA 0 EUR
ABCR20296.0001
3-Ethoxy-2-thenoesäure
3-Ethoxy-2-thenoesäure
Formel: C₇H₈O₃S
Molecular Weight: 172,2 g/mol
MDL: MFCD00205205
CAS-Nummer: 139926-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR