[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid

Lieferant: ABCR

Synonyme: [1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd(dppf)Cl2, 1,1’-Bis(diphenylphosphino)ferrocene palladium(II)dichloride

21063
ABCR21063EA 0 EUR
ABCR21063
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid
Formel: C₃₄H₂₈Cl₂P₂FePd
Molecular Weight: 731,70 g/mol
MDL: MFCD00015757
CAS-Nummer: 72287-26-4
EINECS: 460-040-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR