2-Thiophenboronsäure

Lieferant: ABCR

Synonyme: 2-Thienylboronsäure, Thiophen-2-boronsäure

23071
ABCR23071EA 0 EUR
ABCR23071
2-Thiophenboronsäure
2-Thiopheneboronic acid
Formel: C₄H₅BO₂S
Molecular Weight: 127,96 g/mol
Schmelzpunkt: 146…147 °C
MDL: MFCD00151850
CAS-Nummer: 6165-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR