Ruthenium(III)chlorid, wasserfrei
Lieferant: ABCR
|
Gefahr
|
Synonyme:
Rutheniumtrichlorid
44-5850
ABCR44-5850EA
0
EUR
ABCR44-5850
Ruthenium(III)chlorid, wasserfrei
Ruthenium(III)chlorid
Formel:
Cl₃Ru Molecular Weight: 207,43 g/mol Schmelzpunkt: >500 °C (Zers.) Dichte: 3,11 g/cm³ (20 °C) |
MDL:
MFCD00011208 CAS-Nummer: 10049-08-8 EINECS: 233-167-5 UN: 1759 ADR: 8,II Merck Index: 12,08455 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...