5-Hexin-1-ol

Lieferant: ABCR
Achtung

A11071.0025
ABCRA11071.0025EA 0 EUR
ABCRA11071.0025
5-Hexin-1-ol
5-Hexin-1-ol
Formel: C₆H₁₀O
Molecular Weight: 98,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002980
CAS-Nummer: 928-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR