(±)-β-Aminophenethylalkohol

Lieferant: ABCR

Synonyme: DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol

AA102.0005
ABCRAA102.0005EA 0 EUR
ABCRAA102.0005
(±)-β-Aminophenethylalkohol
(±)-β-Aminophenethylalkohol
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
MDL: MFCD00130145
CAS-Nummer: 7568-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR