Ruthenium(III)chlorid Hydrat

Lieferant: ABCR
Gefahr

Synonyme: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

AB101280.0025
ABCRAB101280.0025EA 0 EUR
ABCRAB101280.0025
Ruthenium(III)chlorid Hydrat
Ruthenium(III)chlorid Hydrat
Formel: Cl₃Ru·xH₂O
Molecular Weight: 207,43 g/mol (anhy)
Schmelzpunkt: > 300 °C
Dichte: 3,11 g/cm³ (20 °C)
MDL: MFCD00149844
CAS-Nummer: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR