10-Undecen-1-ol

Lieferant: ABCR

Synonyme: 1-Undecin-11-ol

AB102120.0025
ABCRAB102120.0025EA 0 EUR
ABCRAB102120.0025
10-Undecen-1-ol
10-Undecin-1-ol
Formel: C₁₁H₂₀O
Molecular Weight: 168,28 g/mol
MDL: MFCD00041675
CAS-Nummer: 2774-84-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR