4-Brom-2-fluorphenol
Lieferant: ABCR
|
Achtung
|
Synonyme:
1-Bromo-3-fluoro-4-hydroxybenzene
AB102153.0250
ABCRAB102153.0250EA
0
EUR
ABCRAB102153.0250
4-Brom-2-fluorphenol
4-Brom-2-fluorphenol
Formel:
C₆H₄BrFO Molecular Weight: 191 g/mol Siedepunkt: 90 °C (15 mmHg) Dichte: 1,74 g/cm³ (20 °C) |
MDL:
MFCD00011722 CAS-Nummer: 2105-94-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...