Dinatriumtetrachloropalladat

Lieferant: ABCR

Synonyme: Natriumtetrachloropalladat(II), Natrium-Palladium(II)-chlorid

AB102748.0005
ABCRAB102748.0005EA 0 EUR
ABCRAB102748.0005
Dinatriumtetrachloropalladat
Dinatriumtetrachloropalladat
Formel: Cl₄Na₂Pd
Molecular Weight: 294,21 g/mol
MDL: MFCD00003487
CAS-Nummer: 13820-53-6
EINECS: 237-502-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR