1H,1H,2H,2H-Perfluoroctyltriethoxysilan

Lieferant: ABCR

Synonyme: Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;

AB104055.0100 AB104055.0005
ABCRAB104055.0100EA 765 EUR
ABCRAB104055.0100 ABCRAB104055.0005
1H,1H,2H,2H-Perfluoroctyltriethoxysilan
1H,1H,2H,2H-Perfluoroctyltriethoxysilan
Formel: C₁₄H₁₉F₁₃O₃Si
Molecular Weight: 510,367 g/mol
MDL: MFCD00042333
CAS-Nummer: 51851-37-7
EINECS: 257-473-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR