2,2,2-Trifluoracetylchlorid

Lieferant: ABCR

AB104142.0100
ABCRAB104142.0100EA 515 EUR
ABCRAB104142.0100
2,2,2-Trifluoracetylchlorid
2,2,2-Trifluoracetylchlorid
Formel: C₂ClF₃O
CAS-Nummer: 354-32-5
EINECS: 206-556-2
UN: 3057
ADR: 2,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR