2,2,2-Trifluorethylacrylat

Lieferant: ABCR

Synonyme: Acrylsäure(2,2,2-trifluorethyl)ester, Propensäure(2,2,2-trifluorethyl)ester

AB104229.0100
ABCRAB104229.0100EA 0 EUR
ABCRAB104229.0100
2,2,2-Trifluorethylacrylat
2,2,2-Trifluorethylacrylat
Formel: C₅H₅F₃O₂
Molecular Weight: 154,09 g/mol
MDL: MFCD00000444
CAS-Nummer: 407-47-6
EINECS: 206-987-6
UN: 3272
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR