Trimethyloxoniumtetrafluoroborat

Lieferant: ABCR

AB104372.0010
ABCRAB104372.0010EA 0 EUR
ABCRAB104372.0010
Trimethyloxoniumtetrafluoroborat
Trimethyloxonium tetrafluoroborate
Formel: C₃H₉BF₄O
Molecular Weight: 147,91 g/mol
MDL: MFCD00011798
CAS-Nummer: 420-37-1
EINECS: 206-994-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR