(Diethylamino)schwefeltrifluorid

Lieferant: ABCR

Synonyme: DAST

AB104704.0100
ABCRAB104704.0100EA 0 EUR
ABCRAB104704.0100
(Diethylamino)schwefeltrifluorid
(Diethylamino)schwefeltrifluorid
Formel: C₄H₁₀F₃NS
Molecular Weight: 161,19 g/mol
Dichte: 1,316 g/cm³ (20 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD00000363
CAS-Nummer: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR