Diphenylphosphin ≥99%

Lieferant: ABCR
Gefahr

AB105242.0010
ABCRAB105242.0010EA 0 EUR
ABCRAB105242.0010
Diphenylphosphin ≥99%
Diphenylphosphin

Phosphine

Formel: C₁₂H₁₁P
Molecular Weight: 186,19 g/mol
Siedepunkt: 116…119 °C (2,6 mmHg)
Dichte: 1,07
MDL: MFCD00003040
CAS-Nummer: 829-85-6
EINECS: 212-591-4
UN: 2845
ADR: 4.2,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR