Bis(cyclopentadienyl)cobalt ≥98%

Lieferant: abcr
Gefahr

Synonyme: Cobaltocene, Bis(cyclopentadienyl)cobalt(II)

AB106088.0001
ABCRAB106088.0001EA 0 EUR
ABCRAB106088.0001
Bis(cyclopentadienyl)cobalt ≥98%
Bis(cyclopentadienyl)cobalt

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formel: C₁₀H₁₀Co
Schmelzpunkt: 173 °C
MDL: MFCD00013749
CAS-Nummer: 1277-43-6
EINECS: 215-061-0
UN: 1325
ADR: 4.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR