Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')europium(III)

Lieferant: ABCR

Synonyme: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)europium(III), Eu(TMHD)3

AB106199.0005
ABCRAB106199.0005EA 0 EUR
ABCRAB106199.0005
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')europium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')europium(III)
Formel: C₃₃H₅₇EuO₆
Molecular Weight: 701,766 g/mol
MDL: MFCD00000019
CAS-Nummer: 15522-71-1
EINECS: 239-565-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR