Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')gadolinium(III)

Lieferant: ABCR

Synonyme: Gd(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)gadolinium(III)

AB106202.0005
ABCRAB106202.0005EA 0 EUR
ABCRAB106202.0005
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')gadolinium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')gadolinium(III)
Formel: C₃₃H₅₇GdO₆
Molecular Weight: 707,052 g/mol
MDL: MFCD00010349
CAS-Nummer: 14768-15-1
EINECS: 238-834-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR