Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)
Lieferant: ABCR
Synonyme:
Y(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)
AB106364.0005
ABCRAB106364.0005EA
0
EUR
ABCRAB106364.0005
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)
Tris(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')yttrium(III)
Formel:
C₃₃H₅₇O₆Y Molecular Weight: 638,708 g/mol |
MDL:
MFCD00015713 CAS-Nummer: 15632-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...