4-Fluor-2-(trifluormethyl)benzaldehyd

Lieferant: ABCR

AB108311.0005
ABCRAB108311.0005EA 0 EUR
ABCRAB108311.0005
4-Fluor-2-(trifluormethyl)benzaldehyd
4-Fluoro-2-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
Siedepunkt: 114 °C (1013 hPa)
Dichte: 1,4 g/cm³ (20 °C)
MDL: MFCD00061313
CAS-Nummer: 90176-80-0
UN: 1989
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR