1-Isopropylpiperazin

Lieferant: abcr

AB108838.0010
ABCRAB108838.0010EA 0 EUR
ABCRAB108838.0010
1-Isopropylpiperazin
1-Isopropylpiperazin
Formel: C₇H₁₆N₂
MDL: MFCD00167971
CAS-Nummer: 4318-42-7
EINECS: 224-345-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR