1,3-Dibrompropan ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Trimethylenbromid

A14687.30 A14687.0B A14687.0I AB109197.1000
A14687.30EA 50.8 EUR
A14687.30 A14687.0B A14687.0I ABCRAB109197.1000
1,3-Dibrompropan ≥98%
1,3-Dibrompropan

Purity: 98% CAS No.: 109-64-8 Molecular Formula: C3H6Br2 Formula Weight: 201.90 Flash Point: 54°(130°F) Physical Form: Liquid Density: 1.99 Refractive Index: 1.5230 Appearance: Colourless Melting Point: -34° Boiling Point: 167° Beilstein Reference: 635662 Merck Reference: 14,9713 UN No.: 1993 MDL No.: MFCD00000255

Formel: BrH₂CCH₂CH₂Br
Molecular Weight: 201.89 g/mol
Siedepunkt: 167 °C
Schmelzpunkt: –34 °C
Dichte: 1.99
Flammpunkt: 54 °C (130 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00000255
CAS-Nummer: 109-64-8
EINECS: 203-690-3
UN: 1993
ADR: 3 III
Merck Index: 12,09844

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR