Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium

Lieferant: abcr

Synonyme: Barium 2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate, Barium(II) 2,2,6,6-tetramethylheptane-3,5-dionate hydrate

AB109919.0005
ABCRAB109919.0005EA 0 EUR
ABCRAB109919.0005
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')barium
Formel: C₂₂H₃₈BaO₄
Molecular Weight: 503.85 g/mol
MDL: MFCD00058709
CAS-Nummer: 17594-47-7
UN: 1564
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR