(3-Aminopropyl)dimethylethoxysilan

Lieferant: abcr

Synonyme: 3-(Ethoxydimethylsilyl)propan-1-amine

AB110423.0025
ABCRAB110423.0025EA 0 EUR
ABCRAB110423.0025
(3-Aminopropyl)dimethylethoxysilan
(3-Aminopropyl)dimethylethoxysilan
Formel: C₇H₁₉NOSi
Molecular Weight: 161.3174 g/mol
MDL: MFCD00053947
CAS-Nummer: 18306-79-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR