L(+)-Arginin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 98+%
CAS No.: 74-79-3
Molecular Formula: C6H14N4O2
Formula Weight: 174.20
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: White
Melting Point: 226° dec.
Beilstein Reference: 1725413
Merck Reference: 14,780
Optical Rotation: +26° (c=1.6 in 6N HCl)
MDL No.: MFCD00002635
Formel:
C₆H₁₄N₄O₂ Siedepunkt: 368 °C (1013 hPa) Schmelzpunkt: 226 °C (Zersetzung) Dichte: 1,3 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002635 CAS-Nummer: 74-79-3 EINECS: 200-811-1 Merck Index: 14,780 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...