(Triphenylphosphin)gold(I)chlorid 98% (99,9% Au) (99.9+%-Au)

Lieferant: abcr
Achtung

Synonyme: Chlorotriphenylphosphine gold(I), Chloro(triphenylphosphine)gold(I), Triphenylphosphinechlorogold

AB110745.0001
ABCRAB110745.0001EA 0 EUR
ABCRAB110745.0001
(Triphenylphosphin)gold(I)chlorid 98% (99,9% Au) (99.9+%-Au)
(Triphenylphosphine)gold(I) chloride
Formel: C₁₈H₁₅AuClP
Molecular Weight: 494.71 g/mol
MDL: MFCD00009588
CAS-Nummer: 14243-64-2
EINECS: 238-117-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR