1,2-Cyclohexandion ≥98%
Lieferant: ABCR
AB110766.0025
ABCRAB110766.0025EA
0
EUR
ABCRAB110766.0025
1,2-Cyclohexandion ≥98%
1,2-Cyclohexandion
Formel:
C₆H₈O₂ Molecular Weight: 112,13 g/mol Siedepunkt: 193…195 °C Schmelzpunkt: 35…38 °C Dichte: 1,12 |
MDL:
MFCD00001648 CAS-Nummer: 765-87-7 EINECS: 212-155-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...