(3-Brompropyl)trimethoxysilan
Lieferant: ABCR
Synonyme:
Trimethoxy(3-bromopropyl)silane, 3-(Trimethoxysilyl)propylbromide
AB110909.0010
ABCRAB110909.0010EA
0
EUR
ABCRAB110909.0010
(3-Brompropyl)trimethoxysilan
(3-Brompropyl)trimethoxysilan
Formel:
BrH₂CCH₂CH₂Si(OCH₃)₃ Molecular Weight: 243,17 g/mol |
MDL:
MFCD00053894 CAS-Nummer: 51826-90-5 EINECS: 257-453-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...