tert-Butyldimethylsilyltrifluormethansulfonat

Lieferant: ABCR

Synonyme: t-Butyldimethylsilyl trifluoromethanesulphonate

AB110921.0010
ABCRAB110921.0010EA 0 EUR
ABCRAB110921.0010
tert-Butyldimethylsilyltrifluormethansulfonat
tert-Butyldimethylsilyltrifluormethansulfonat
Formel: C₇H₁₅F₃O₃SSi
Molecular Weight: 264,34 g/mol
MDL: MFCD00000405
CAS-Nummer: 69739-34-0
EINECS: 274-102-0
UN: 2920
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR