(3-Chlorpropyl)triethoxysilan

Lieferant: ABCR

Synonyme: 3-(Triethoxysilyl)propylchlorid, Triethoxy(3-chlorpropyl)silan

AB110982.0025
ABCRAB110982.0025EA 0 EUR
ABCRAB110982.0025
(3-Chlorpropyl)triethoxysilan
(3-Chlorpropyl)triethoxysilan
Formel: ClH₂CCH₂CH₂Si(OCH₂CH₃)₃
Molecular Weight: 240,8 g/mol
MDL: MFCD00018985
CAS-Nummer: 5089-70-3
EINECS: 225-805-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR