(Dichlormethyl)dimethylchlorsilan

Lieferant: ABCR

AB111035.0025
ABCRAB111035.0025EA 0 EUR
ABCRAB111035.0025
(Dichlormethyl)dimethylchlorsilan
(Dichlormethyl)dimethylchlorsilan
Formel: C₃H₇Cl₃Si
MDL: MFCD00018852
CAS-Nummer: 18171-59-0
EINECS: 242-058-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR