Diethoxy(dimethyl)silan

Lieferant: ABCR

Synonyme: Dimethyldiethoxysilan

SID4121.0 AB111080.0100
ABCRSID4121.0EA 0 EUR
ABCRSID4121.0 ABCRAB111080.0100
Diethoxy(dimethyl)silan
Diethoxy(dimethyl)silan
Formel: C₆H₁₆O₂Si
Molecular Weight: 148,28 g/mol
Siedepunkt: 114 °C (1013 hPa)
Schmelzpunkt: –87 °C
Dichte: 0,853 g/cm³ (20 °C)
MDL: MFCD00009068
CAS-Nummer: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR