Hexamethylcyclotrisiloxan
Lieferant: ABCR
Synonyme:
2,2,4,4,6,6-Hexamethyl-1,3,5,2,4,6-trioxatrisilinane
AB111171.0500
ABCRAB111171.0500EA
0
EUR
ABCRAB111171.0500
Hexamethylcyclotrisiloxan
Hexamethylcyclotrisiloxane
Formel:
C₆H₁₈O₃Si₃ Molecular Weight: 222,46 g/mol Siedepunkt: 134 °C (1013 hPa) Schmelzpunkt: 64,5 °C Dichte: 1,12 g/cm³ (20 °C) |
MDL:
MFCD00005943 CAS-Nummer: 541-05-9 EINECS: 208-765-4 UN: 1325 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...