Phenyltrimethoxysilan

Lieferant: abcr

AB111342.0100
ABCRAB111342.0100EA 0 EUR
ABCRAB111342.0100
Phenyltrimethoxysilan
Phenyltrimethoxysilan
Formel: C₉H₁₄O₃Si
Dichte: 1,064 g/cm³ (20 °C)
MDL: MFCD00025689
CAS-Nummer: 2996-92-1
EINECS: 221-066-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR