1,3-Dichlor-1,1,3,3-tetraisopropyldisiloxan

Lieferant: abcr

Synonyme: 1,3-Dichloro-1,1,3,3-tetrakis(isopropyl)disiloxane

AB111393.0025
ABCRAB111393.0025EA 0 EUR
ABCRAB111393.0025
1,3-Dichlor-1,1,3,3-tetraisopropyldisiloxan
1,3-Dichlor-1,1,3,3-tetraisopropyldisiloxan
Formel: C₁₂H₂₈Cl₂OSi₂
Molecular Weight: 315.43 g/mol
MDL: MFCD00009655
CAS-Nummer: 69304-37-6
UN: 1760
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR