1H,1H,2H,2H-Perfluoroctyltrichlorsilan

Lieferant: ABCR

Synonyme: Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-trichlorosilane

AB111444.0010
ABCRAB111444.0010EA 140 EUR
ABCRAB111444.0010
1H,1H,2H,2H-Perfluoroctyltrichlorsilan
1H,1H,2H,2H-Perfluoroctyltrichlorsilan
Formel: C₈H₄Cl₃F₁₃Si
Molecular Weight: 481,55 g/mol
MDL: MFCD00042363
CAS-Nummer: 78560-45-9
EINECS: 278-947-6
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR