(Trifluormethyl)trimethylsilan

Lieferant: abcr

Synonyme: trimethyl(trifluoromethyl)silane

AB111468.0100
ABCRAB111468.0100EA 0 EUR
ABCRAB111468.0100
(Trifluormethyl)trimethylsilan
(Trifluoromethyl)trimethylsilane
Formel: C₄H₉F₃Si
Siedepunkt: 55,5 °C (1013 hPa)
Dichte: 0,95 g/cm³ (20 °C)
MDL: MFCD00145454
CAS-Nummer: 81290-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR