Tetrapentylzinn

Lieferant: abcr

AB111632.0005
ABCRAB111632.0005EA 0 EUR
ABCRAB111632.0005
Tetrapentylzinn
Tetrapentylzinn
Formel: C₂₀H₄₄Sn
Molecular Weight: 403.28 g/mol
CAS-Nummer: 3765-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR