2,4,6-Trifluorbenzonitril

Lieferant: ABCR

AB112764.0010
ABCRAB112764.0010EA 0 EUR
ABCRAB112764.0010
2,4,6-Trifluorbenzonitril
2,4,6-Trifluorbenzonitril
Formel: C₇H₂F₃N
Molecular Weight: 157,09 g/mol
Siedepunkt: 179 °C (1013 hPa)
Schmelzpunkt: 57…58 °C
MDL: MFCD00042399
CAS-Nummer: 96606-37-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR