Tris(triphenylphosphin)rhodium(I)chlorid ≥99%
Lieferant: abcr
|
Achtung
|
AB113017.0005
ABCRAB113017.0005EA
0
EUR
ABCRAB113017.0005
Tris(triphenylphosphin)rhodium(I)chlorid ≥99%
Tris(triphenylphosphin)rhodium(I)chlorid
Formel:
C₅₄H₄₅ClP₃Rh Schmelzpunkt: 157 °C Dichte: 1,363 g/cm³ (20 °C) |
MDL:
MFCD00010016 CAS-Nummer: 14694-95-2 EINECS: 238-744-5 Merck Index: 12,02226 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...