N(α)-Boc-L-tryptophan ≥98%
Lieferant: abcr
AB114355.0005
ABCRAB114355.0005EA
0
EUR
ABCRAB114355.0005
N(α)-Boc-L-tryptophan ≥98%
N(α)-Boc-L-tryptophan
Purity: 98+%
CAS No.: 13139-14-5
Molecular Formula: C16H20N2O4
Formula Weight: 304.35
Storage Temperature: 2°C to 8°C
Melting Point: ca 138° dec.
Beilstein Reference: 39677
Optical Rotation: -19.5° (c=1 in DMF)
MDL No.: MFCD00065595
Formel:
C₁₆H₂₀N₂O₄ Lagertemperatur: Kühlschrank |
MDL:
MFCD00065595 CAS-Nummer: 13139-14-5 EINECS: 236-072-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...