2-(Dimethylamino)benzoesäure 98%

Lieferant: ABCR
Achtung

Synonyme: 2-Dimethylaminobenzoic acid

AB116140.0001
ABCRAB116140.0001EA 0 EUR
ABCRAB116140.0001
2-(Dimethylamino)benzoesäure 98%
2-(Dimethylamino)benzoesäure

Purity: 98% CAS No.: 610-16-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Odour: Odourless Melting Point: 70-74° Beilstein Reference: 2089466 MDL No.: MFCD00016496

Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Schmelzpunkt: 70…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016496
CAS-Nummer: 610-16-2
EINECS: 210-209-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR