Ethyl-trans-3-ethoxyacrylat ≥98%

Lieferant: abcr

AB116420.0250
ABCRAB116420.0250EA 0 EUR
ABCRAB116420.0250
Ethyl-trans-3-ethoxyacrylat ≥98%
Ethyl-trans-3-ethoxyacrylat
Formel: C₇H₁₂O₃
MDL: MFCD00009863
CAS-Nummer: 5941-55-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR